About This Item
mp
148-151 °C (lit.)
storage temp.
−20°C
SMILES string
Cl[H].[H][C@]1(CCCN1)C(=O)OCc2ccccc2
InChI
1S/C12H15NO2.ClH/c14-12(11-7-4-8-13-11)15-9-10-5-2-1-3-6-10;/h1-3,5-6,11,13H,4,7-9H2;1H/t11-;/m0./s1
InChI key
NEDMOHHWRPHBAL-MERQFXBCSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service