Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8625

Sigma-Aldrich

Perylene

≥98%

Synonym(s):

Dibenz[de,kl]anthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein/REAXYS Number:
1911335
EC Number:
MDL number:
UNSPSC Code:
12171500

assay

≥98%

mp

276-279 °C (lit.)

SMILES string

c1cc2cccc3c4cccc5cccc(c(c1)c23)c45

InChI

1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H

InChI key

CSHWQDPOILHKBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service