P8449
L-Proline
BioXtra, pH 6.0-7.0 (20 °C, 1 M in H2O), >99% (TLC)
Synonym(s):
(S)-Pyrrolidine-2-carboxylic acid
About This Item
Recommended Products
product line
BioXtra
assay
>99% (TLC)
impurities
<0.0005% Phosphorus (P)
<0.1% Insoluble matter
ign. residue
<0.1%
pH
6.0-7.0 (20 °C, 1 M in H2O)
mp
228 °C (dec.) (lit.)
solubility
H2O: 1 M at 20 °C, clear, colorless
anion traces
chloride (Cl-): <0.05%
sulfate (SO42-): <0.05%
cation traces
Al: <0.0005%
Ca: <0.0005%
Cu: <0.0005%
Fe: <0.0005%
K: <0.005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.005%
Pb: <0.001%
Zn: <0.0005%
absorption
<0.05 at 260 in H2O at 1 M
<0.05 at 280 in H2O at 1 M
SMILES string
OC(=O)[C@@H]1CCCN1
InChI
1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1
InChI key
ONIBWKKTOPOVIA-BYPYZUCNSA-N
Looking for similar products? Visit Product Comparison Guide
Application
replaced by
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service