Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P5802

Sigma-Aldrich

Potassium peroxodisulfate

suitable for electrophoresis, suitable for catalyst of acrylamide polymerization, >99%

Synonym(s):

Potassium persulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2S2O8
CAS Number:
Molecular Weight:
270.32
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

>99%

reaction suitability

reagent type: oxidant

technique(s)

electrophoresis: suitable

impurities

≤0.005% Insoluble matter

anion traces

chloride (Cl-): ≤0.001%

cation traces

Fe: ≤5 ppm

suitability

suitable for catalyst of acrylamide polymerization

SMILES string

[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O

InChI

1S/2K.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

USHAGKDGDHPEEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernaurdshaw Neppolian et al.
Water research, 44(12), 3687-3695 (2010-04-27)
The sonochemical oxidation of As(III) in the presence of peroxydisulfate ion (PDS) has been investigated. Sulfate anion radicals and OH radicals produced during acoustic cavitation readily oxidized As(III) to As(V) in an aqueous environment. The rate of oxidation of As(III)
Huan Niu et al.
Biosensors & bioelectronics, 26(7), 3175-3180 (2011-01-18)
A novel label free electrochemiluminescence (ECL) immunosensor based on the ECL of peroxydisulfate solution for detection of α-1-fetoprotein (AFP) has been developed. For this proposed immunosensor, L-cysteine was firstly electrodeposited on the gold electrode surface, which promoted the electron transfer
Harriet Mörtstedt et al.
Rapid communications in mass spectrometry : RCM, 25(2), 327-340 (2010-12-31)
Oxidative stress has been suggested as an underlying mechanism of many human diseases. However, definitive evidence for this association has not been presented due to different shortcomings of the methods used to measure biomarkers of oxidative stress. Persulfates are oxidizing
Zhiyong Yang et al.
The Journal of organic chemistry, 77(16), 7086-7091 (2012-07-28)
Nontransition metal-catalyzed synthesis of 2-aryl benzothiazoles was achieved through K(2)S(2)O(8)-mediated oxidative condensation of benzothiazoles with aryl aldehydes. The same transformation can also be effected when the aryl aldehydes were replaced with phenylglyoxylic acids.
Xiuping Zhu et al.
Environmental science & technology, 48(12), 7157-7163 (2014-05-28)
Several technologies, including pressure-retarded osmosis (PRO), reverse electrodialysis (RED), and capacitive mixing (CapMix), are being developed to recover energy from salinity gradients. Here, we present a new approach to capture salinity gradient energy based on the expansion and contraction properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service