P4273
1,2-Diheptanoyl-sn-glycero-3-phosphocholine
solution, 20 mg/mL in chloroform, ~99% (GC)
Synonym(s):
L-α-Phosphatidylcholine, diheptanoyl
About This Item
Recommended Products
assay
~99% (GC)
form
solution
concentration
20 mg/mL in chloroform
storage temp.
−20°C
SMILES string
CCCCCCC(=O)OC[C@@H](COP(O)(=O)OCC[N](C)(C)C)OC(=O)CCCCCC
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
target_organs
Central nervous system, Liver,Kidney
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service