Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4173

Supelco

(+)-Pseudoephedrine hydrochloride solution

1.0 mg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO · HCl
CAS Number:
Molecular Weight:
201.69
EC Number:
MDL number:
UNSPSC Code:
41116107

availability

not available in Japan

concentration

1.0 mg/mL in methanol

storage temp.

2-8°C

SMILES string

Cl.CN[C@H](C)[C@H](O)c1ccccc1

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service