Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3280

Supelco

D-Pantethine hydrate

syrup

Synonym(s):

Bis(N-pantothenylamidoethyl) disulfide, LBF factor, Pantethein oxidized form

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H42N4O8S2 · xH2O
CAS Number:
Molecular Weight:
554.72 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

form

syrup

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

~20% water

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

InChI

1S/C22H42N4O8S2/c1-21(2,13-27)17(31)19(33)25-7-5-15(29)23-9-11-35-36-12-10-24-16(30)6-8-26-20(34)18(32)22(3,4)14-28/h17-18,27-28,31-32H,5-14H2,1-4H3,(H,23,29)(H,24,30)(H,25,33)(H,26,34)/t17-,18-/m0/s1

InChI key

DJWYOLJPSHDSAL-ROUUACIJSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Comparable product

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Webster
European journal of applied physiology and occupational physiology, 77(6), 486-491 (1998-07-03)
The purpose of this study was to investigate the physiological and performance responses to supplementation with allithiamin and pantethine. On two separate occasions, six highly trained cyclists [maximum O2 consumption or VO2max 61.8 (2.1) ml x kg(-1) x min(-1)] performed
Mark Houston
Journal of clinical hypertension (Greenwich, Conn.), 14(2), 121-132 (2012-01-27)
The combination of a lipid-lowering diet and scientifically proven nutraceutical supplements has the ability to significantly reduce low-density lipoprotein (LDL) cholesterol, increase LDL particle size, decrease LDL particle number, lower trigylcerides and very LDL levels, and increase total and high-density
J J Harding
Drugs & aging, 18(7), 473-486 (2001-08-03)
Cataract is the major cause of blindness and of visual impairment worldwide, so its prevention is of the greatest importance. At present no drug therapy is licensed for use in the UK or the US, so the only treatment for
J I Clark et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(26), 15185-15189 (1996-12-24)
The effects of pantethine, glutathione, and selected chemical reagents on the anti-aggregation activity of alpha-crystallin was evaluated. Protein aggregation was monitored by light scattering of solutions of denatured beta L-crystallin or alcohol dehydrogenase (ADH). The ratios of beta L-crystallin/alpha-crystallin and
Carl J Balibar et al.
Journal of bacteriology, 193(13), 3304-3312 (2011-05-10)
Coenzyme A (CoA) plays a central and essential role in all living organisms. The pathway leading to CoA biosynthesis has been considered an attractive target for developing new antimicrobial agents with novel mechanisms of action. By using an arabinose-regulated expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service