Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2426

Sigma-Aldrich

D-Pyroglutamic acid

Synonym(s):

(R)-2-Pyrrolidone-5-carboxylic acid, (R)-5-Oxo-2-pyrrolidinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
Beilstein/REAXYS Number:
82133
EC Number:
MDL number:
UNSPSC Code:
12352200

SMILES string

OC(=O)[C@H]1CCC(=O)N1

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service