Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2375

Sigma-Aldrich

D-Panthenol

≥98% (perchloric acid titration), ≥98% (TLC)

Synonym(s):

(R)-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, (R)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, D-Pantothenyl alcohol, Dexpanthenol, Provitamin B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO4
CAS Number:
Molecular Weight:
205.25
Beilstein/REAXYS Number:
1724947
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

synthetic (organic)

assay

≥98% (TLC)
≥98% (perchloric acid titration)

form

liquid
semisolid

color

colorless to white

density

1.20 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCCO

InChI

1S/C9H19NO4/c1-9(2,6-12)7(13)8(14)10-4-3-5-11/h7,11-13H,3-6H2,1-2H3,(H,10,14)/t7-/m0/s1

InChI key

SNPLKNRPJHDVJA-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Göbbels et al.
Klinische Monatsblatter fur Augenheilkunde, 209(2-3), 84-88 (1996-08-01)
In this controlled, randomized, double-masked study the effect of dexpanthenol-containing artificial tears (Siccaprotect) on patients with dry eyes was examined. 50 patients applied either dexpanthenol-containing artificial tears (Siccaprotect) or the identical, but free of dexpanthenol, eye drops five times daily
M Sachs et al.
Zeitschrift fur Ernahrungswissenschaft, 29(4), 270-283 (1990-12-01)
The aim of this study was the examination of the metabolism and mechanism of action of D-pantothenyl alcohol in patients with postoperative intestinal atony. Seven metabolically healthy patients were examined on the 4th day following colorectal surgery, before bowel activity
Fritz Ebner et al.
American journal of clinical dermatology, 3(6), 427-433 (2002-07-13)
Pantothenic acid is essential to normal epithelial function. It is a component of coenzyme A, which serves as a cofactor for a variety of enzyme-catalyzed reactions that are important in the metabolism of carbohydrates, fatty acids, proteins, gluconeogenesis, sterols, steroid
W Gehring et al.
Arzneimittel-Forschung, 50(7), 659-663 (2000-08-31)
In a randomized, double-blind, placebo-controlled study the effect of topical dexpanthenol (CAS 81-13-0) formulated in two different lipophilic vehicles on epidermal barrier function in vivo was carried out. Seven days' treatment with dexpanthenol improved stratum corneum hydration and reduced transepidermal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service