P103
(S)-(−)-3-(3-Hydroxyphenyl)-N-propylpiperidine hydrochloride
≥98% (HPLC), solid
Synonym(s):
Preclamol, S-(−)-3-PPP
About This Item
Recommended Products
assay
≥98% (HPLC)
form
solid
color
white
solubility
H2O: 150 mg/mL (Solutions should be freshly prepared.)
ethanol: slightly soluble (Solutions should be freshly prepared.)
SMILES string
Cl.CCCN1CCC[C@H](C1)c2cccc(O)c2
InChI
1S/C14H21NO.ClH/c1-2-8-15-9-4-6-13(11-15)12-5-3-7-14(16)10-12;/h3,5,7,10,13,16H,2,4,6,8-9,11H2,1H3;1H/t13-;/m1./s1
InChI key
NRHUDETYKUBQJT-BTQNPOSSSA-N
Gene Information
human ... DRD1(1812) , DRD2(1813) , DRD3(1814) , DRD4(1815)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service