P0704
1-O-Palmityl-2-(cis-8,11,14-eicosatrienoyl)-sn-glycero-3-phosphocholine
~25 mg/mL in ethanol
Synonym(s):
L-α-Phosphatidylcholine, β-(cis-8,11,14-eicosatrienoyl)-γ-O-hexadecyl
About This Item
concentration
~25 mg/mL in ethanol
functional group
phospholipid
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCOC[C@@H](COP(O)(=O)OCC[N](C)(C)C)OC(=O)CCCCCC\C=C/C\C=C/C\C=C/CCCCC
InChI
1S/C44H85NO7P/c1-6-8-10-12-14-16-18-20-22-23-24-25-27-29-31-33-35-37-44(46)52-43(42-51-53(47,48)50-40-38-45(3,4)5)41-49-39-36-34-32-30-28-26-21-19-17-15-13-11-9-7-2/h14,16,20,22,24-25,43H,6-13,15,17-19,21,23,26-42H2,1-5H3,(H,47,48)/b16-14-,22-20-,25-24-
InChI key
BRXGGXRZUYJPTM-GBDFTIALSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
55.4 °F
flash_point_c
13 °C
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service