Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0251

Sigma-Aldrich

Phenolphthalein disulfate tripotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H13O11S2K3
CAS Number:
Molecular Weight:
646.77
MDL number:
UNSPSC Code:
12171500

storage temp.

−20°C

SMILES string

[K+].[K+].[K+].OC(c1ccc(OS([O-])(=O)=O)cc1)(c2ccc(OS([O-])(=O)=O)cc2)c3ccccc3C([O-])=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service