Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0152

Sigma-Aldrich

Prednisolone

analytical standard

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-3,20-dione, 1-Dehydrocortisol, 1-Dehydrohydrocortisone, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O5
CAS Number:
Molecular Weight:
360.44
Beilstein/REAXYS Number:
1354103
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

packaging

vial of 250 mg

impurities

≤1.5 mol/mol water

mp

240 °C (dec.) (lit.)

SMILES string

O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2([H])[C@@H](O)C[C@@]4(C)[C@@]3([H])CC[C@]4(O)C(CO)=O)=C1

InChI

1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

OIGNJSKKLXVSLS-VWUMJDOOSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in amber screw-cap vials

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minoru Ishikawa et al.
Bioorganic & medicinal chemistry letters, 19(5), 1457-1460 (2009-01-31)
A natural product, pseurotin A inhibits IgE production in vitro. Wide variety of chemical modification of pseurotin A was performed. Structure-activity relationship studies of pseurotin analogues elucidated that 10-deoxypseurotin A strongly inhibits IgE production with IC(50) of 0.066 microM. An
Uri Elbaz et al.
Cornea, 34(7), 773-777 (2015-03-27)
To report the outcomes of corneal fine needle diathermy (FND) with adjuvant intrastromal and subconjunctival bevacizumab injection for corneal neovascularization (CN) in children. Medical records of all children who had undergone FND with adjuvant bevacizumab injection were reviewed retrospectively. Treatment
Vappu Rantalaiho et al.
Annals of the rheumatic diseases, 73(11), 1954-1961 (2013-08-03)
To study whether adding initial infliximab to remission-targeted initial combination-DMARD treatment improves the long-term outcomes in patients with early rheumatoid arthritis (RA). Ninety-nine patients with early, DMARD-naïve RA were treated with a triple combination of DMARDs, starting with methotrexate (max
Axel Heidenreich et al.
European journal of cancer (Oxford, England : 1990), 50(6), 1090-1099 (2014-02-04)
Cabazitaxel/prednisone has been shown to prolong survival versus mitoxantrone/prednisone in patients with metastatic castration-resistant prostate cancer (mCRPC) that has progressed during or after docetaxel. Subsequently, compassionate-use programmes (CUPs) and expanded-access programmes (EAPs) were established worldwide, allowing access to cabazitaxel before
J A P Spijkers-Hagelstein et al.
Leukemia, 28(4), 761-769 (2013-08-21)
Successful treatment results for MLL-rearranged Acute Lymphoblastic Leukemia (ALL) in infants remain difficult to achieve. Significantly contributing to therapy failure is poor response to glucocorticoids (GCs), like prednisone. Thus, overcoming resistance to these drugs may be a crucial step towards

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service