Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P0024

Sigma-Aldrich

PG-01

≥98% (HPLC)

Synonym(s):

2-[(2-1H-indol-3-yl-acetyl)-methylamino]-N-(4-isopropylphenyl)-2-phenylacetamide, N-Methyl-N-[2-[[4-(1-methylethyl)phenyl]amino]-2-oxo-1-phenylethyl]-1H-indole-3-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29N3O2
CAS Number:
Molecular Weight:
439.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

−20°C

SMILES string

CC(C)c1ccc(NC(=O)C(N(C)C(=O)Cc2c[nH]c3ccccc23)c4ccccc4)cc1

InChI

1S/C28H29N3O2/c1-19(2)20-13-15-23(16-14-20)30-28(33)27(21-9-5-4-6-10-21)31(3)26(32)17-22-18-29-25-12-8-7-11-24(22)25/h4-16,18-19,27,29H,17H2,1-3H3,(H,30,33)

InChI key

PQAYCXMQTUEDRD-UHFFFAOYSA-N

Biochem/physiol Actions

CFTR mutations are responsible for cystic fibrosis. The most common mutation is ΔF508, but many others exist. Drugs which can correct channel function of a broad range of CFTR mutants are most desirable for clinical development. PG-01 is a potentiator of ΔF508 (Ka 70 nM) as well as mutants G551D and G1349D (Ka 1100 and 40 nM, respectively). Its broad spectrum may make it more desirable than more mutant-specific CFTR correctors.
PG-01 is a potentiator of ΔF508 (Ka 70 nM) as well as mutants G551D and G1349D (Ka 1100 and 40 nM, respectively).

Features and Benefits

This compound is featured on the Chloride Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to chloride channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service