Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O1756

Sigma-Aldrich

Orotic acid

suitable for plant cell culture

Synonym(s):

2,6-Dihydroxypyrimidine-4-carboxylic acid, 6-Carboxy-2,4-dihydroxypyrimidine, 6-Carboxyuracil, Uracil-6-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N2O4
CAS Number:
Molecular Weight:
156.10
Beilstein/REAXYS Number:
383901
EC Number:
MDL number:
UNSPSC Code:
41106305

technique(s)

cell culture | plant: suitable

mp

≥300 °C

SMILES string

OC(=O)C1=CC(=O)NC(=O)N1

InChI

1S/C5H4N2O4/c8-3-1-2(4(9)10)6-5(11)7-3/h1H,(H,9,10)(H2,6,7,8,11)

InChI key

PXQPEWDEAKTCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliana Cheleski et al.
European journal of medicinal chemistry, 45(12), 5899-5909 (2010-10-23)
The enzyme dihydroorotate dehydrogenase (DHODH) has been suggested as a promising target for the design of trypanocidal agents. We report here the discovery of novel inhibitors of Trypanosoma cruzi DHODH identified by a combination of virtual screening and ITC methods.
Costantino Salerno et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 781(1-2), 57-71 (2002-11-27)
Urinary orotic acid determination is a useful tool for screening hereditary orotic aciduria and for differentiating the hyperammonemia disorders which cannot be readily diagnosed by amino acid chromatography, thus reducing the need for enzyme determination in tissue biopsies. This review
Lili Wang et al.
Molecular genetics and metabolism, 105(2), 203-211 (2011-12-03)
Ornithine transcarbamylase deficiency (OTCD), the most common and severe urea cycle disorder, is an excellent model for developing liver-directed gene therapy. No curative therapy exists except for liver transplantation which is limited by available donors and carries significant risk of
Hirokazu Suzuki et al.
Applied and environmental microbiology, 78(20), 7376-7383 (2012-08-14)
Counterselection systems facilitate marker-free genetic modifications in microbes by enabling positive selections for both the introduction of a marker gene into the microbe and elimination of the marker from the microbe. Here we report a counterselection system for Geobacillus kaustophilus
[Orotic acid].
M Falk
Die Pharmazie, 40(6), 377-383 (1985-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service