All Photos(1)
About This Item
Linear Formula:
H2N[CH2]2CT2CT[NH2]CO2H
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
aqueous ethanol solution
extent of labeling
40-70 Ci/mmol
storage temp.
2-8°C
SMILES string
NCCCC(N)C(O)=O
Packaging
serum bottle
Physical form
Aqueous solution containing 2% ethanol
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sarah C Huen et al.
Pediatric nephrology (Berlin, Germany), 30(2), 199-209 (2014-01-21)
Acute ischemic kidney injury is a common complication in hospitalized patients. No treatment is yet available for augmenting kidney repair or preventing progressive kidney fibrosis. Animal models of acute kidney injury demonstrate that activation of the innate immune system plays
Mohammad Ronaghi et al.
Stem cells and development, 23(11), 1275-1284 (2014-02-12)
In mammals, the permanence of many forms of hearing loss is the result of the inner ear's inability to replace lost sensory hair cells. Here, we apply a differentiation strategy to guide human embryonic stem cells (hESCs) into cells of
Marta Biagioli et al.
Human molecular genetics, 24(9), 2442-2457 (2015-01-13)
The CAG repeat expansion in the Huntington's disease gene HTT extends a polyglutamine tract in mutant huntingtin that enhances its ability to facilitate polycomb repressive complex 2 (PRC2). To gain insight into this dominant gain of function, we mapped histone
Maja Sabol et al.
PloS one, 9(12), e114510-e114510 (2014-12-17)
Hedgehog-Gli (Hh-Gli) signaling pathway is one of the new molecular targets found upregulated in breast tumors. Estrogen receptor alpha (ERα) signaling has a key role in the development of hormone-dependent breast cancer. We aimed to investigate the effects of inhibiting
Taito Matsuda et al.
Nature communications, 6, 6514-6514 (2015-03-10)
Pathological conditions such as epilepsy cause misregulation of adult neural stem/progenitor populations in the adult hippocampus in mice, and the resulting abnormal neurogenesis leads to impairment in learning and memory. However, how animals cope with abnormal neurogenesis remains unknown. Here
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service