Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N9883

Sigma-Aldrich

1-Naphthyl isothiocyanate

>95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NCS
CAS Number:
Molecular Weight:
185.24
EC Number:
MDL number:
UNSPSC Code:
12171500

assay

>95%

mp

55.5-57 °C (lit.)

storage temp.

2-8°C

SMILES string

S=C=Nc1cccc2ccccc12

InChI

1S/C11H7NS/c13-8-12-11-7-3-5-9-4-1-2-6-10(9)11/h1-7H

InChI key

JBDOSUUXMYMWQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masayuki Kimura et al.
Journal of applied toxicology : JAT, 36(2), 223-237 (2015-05-27)
We have previously reported that a 28-day treatment of carcinogens evoking target cell proliferation activates G1 /S checkpoint function and apoptosis, as well as induction of aberrant ubiquitin D (Ubd) expression, suggesting disruptive spindle checkpoint function, in rats. The present
Hiroyuki Oka et al.
Toxicology, 404-405, 68-75 (2018-05-20)
The genotoxic potential of drugs is a serious problem, and its evaluation is one of the most critical processes of drug development. Although the comet assay of compound-exposed tissue is a frequently used genotoxicity test, its high false-positive rate is
Dennis J Pelletier et al.
Journal of chemical information and modeling, 47(3), 1196-1205 (2007-04-13)
The identification of phospholipidosis (PPL) during preclinical testing in animals is a recognized problem in the pharmaceutical industry. Depending on the intended indication and dosing regimen, PPL can delay or stop development of a compound in the drug discovery process.
Takeki Uehara et al.
Molecular nutrition & food research, 54(2), 218-227 (2009-12-31)
Biotechnology advances have provided novel methods for the risk assessment of chemicals. The application of microarray technologies to toxicology, known as toxicogenomics, is becoming an accepted approach for identifying chemicals with potential safety problems. Gene expression profiling is expected to
Sean Ekins et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(12), 2302-2308 (2010-09-17)
Drug-induced liver injury (DILI) is one of the most important reasons for drug development failure at both preapproval and postapproval stages. There has been increased interest in developing predictive in vivo, in vitro, and in silico models to identify compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service