Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N6633

Sigma-Aldrich

19-Nortestosterone 17-decanoate

powder

Synonym(s):

17β-Hydroxy-19-nor-4-androsten-3-one 17-decanoate, 17β-Hydroxy-4-estren-3-one 17-decanoate, 4-Estren-17β-ol-3-one 17-decanoate, Decadurabolin, Nandrolone decanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O3
CAS Number:
Molecular Weight:
428.65
EC Number:
MDL number:
UNSPSC Code:
51111800

form

powder

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to light yellow

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.8 mg/mL
DMSO: 10 mg/mL
ethanol: 5 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C

InChI

1S/C28H44O3/c1-3-4-5-6-7-8-9-10-27(30)31-26-16-15-25-24-13-11-20-19-21(29)12-14-22(20)23(24)17-18-28(25,26)2/h19,22-26H,3-18H2,1-2H3/t22-,23+,24+,25-,26-,28-/m0/s1

InChI key

JKWKMORAXJQQSR-MOPIKTETSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anabolic steroid with a stimulatory effect on bone mass, muscle ultrastructure, and immune response.

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena L Gomes et al.
Canadian journal of physiology and pharmacology, 90(1), 75-82 (2012-01-06)
Several studies have demonstrated that gonadal hormones show significant effects on the brain and signaling pathways of effector organs/cells that respond to neurotransmitters. Since little information is available concerning the impact of male and female gonadal hormones on the renal
Rita Cássia Marqueti et al.
European journal of applied physiology, 112(4), 1505-1515 (2011-08-16)
The aim of this study was to evaluate the expression of key genes responsible for tendon remodeling of the proximal and distal regions of calcaneal tendon (CT), intermediate and distal region of superficial flexor tendon (SFT) and proximal, intermediate and
Carolina Almeida do Carmo et al.
Journal of applied toxicology : JAT, 32(10), 810-814 (2011-07-01)
Nandrolone is an androgenic-anabolic steroid (AAS) with diverse medical applications but taken indiscriminately by some to rapidly increase muscle mass. The aim of this study was to evaluate the genotoxic and clastogenic potential of nandrolone (deca-durabolin®) in vivo in different
Gustavo Tralli Nogueira et al.
Lasers in medical science, 27(6), 1189-1193 (2011-12-23)
There has recently been constant effort to evaluate therapies that may have a positive effect on bone regeneration. However, there are few studies in the literature on the effects of low-level laser therapy (LLLT) involving tissues treated with anabolic steroids.
Fernanda Carvalho Cavalari et al.
Steroids, 77(1-2), 118-125 (2011-11-19)
Studies show a mechanism of action of testosterone, nandrolone and catechin as agonists of the membrane androgen receptor. The aim of this work is to investigate the non-classical effect of androgens and catechin in Sertoli cells from immature rats. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service