Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N6007

Sigma-Aldrich

1-Nitrosopiperidine

Synonym(s):

NPIP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O
CAS Number:
Molecular Weight:
114.15
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

density

1.06 g/mL (lit.)

storage temp.

2-8°C

SMILES string

O=NN1CCCCC1

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Carcinogen that induces benign and malignant tumors of the respiratory tract, stomach and esophagus in rodents; potent nasal carcinogen in rats.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel González-Mancebo et al.
Mutation research, 558(1-2), 45-51 (2004-03-24)
The genotoxicity of nitrosopiperidine and six alkyl derivatives was studied by use of the Ames tester strains TA100 and TA1535 in the pre-incubation method. Among the compounds investigated, those exhibiting genotoxic activity under the experimental conditions employed were only genotoxic
P Levallois et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 38(11), 1013-1019 (2000-10-19)
Urinary excretion of volatile nitrosamines was assessed in 59 non-smokers living in a rural county of Québec, Canada. Water and food intakes were measured by means of a 24-hour recall. Nitrates were analyzed in the tap water of all participants
Y Yamashita et al.
Mutation research, 348(4), 163-168 (1995-12-01)
N-Nitrosodialkylamines are promutagens and proclastogens, requiring metabolic activation for their actions. Previously, we showed that direct-acting bacterial mutagens can be formed from N-nitrosodialkylamines on exposure to near-UV. We have now found that N-nitrosodialkylamines with near-UV irradiation are clastogenic to Chinese
Dennie G A J Hebels et al.
Toxicology letters, 207(3), 232-241 (2011-09-29)
N-nitroso compounds (NOCs) may represent a carcinogenic risk to humans following endogenous colonic nitrosation processes. We used the colon adenocarcinoma cell line Caco-2 to investigate transcriptomic changes at three time points (1, 6, 24 h) following exposure to genotoxic concentrations
I T Vermeer et al.
Mutation research, 428(1-2), 353-361 (1999-10-13)
Many constituents present in the human diet may inhibit endogenous formation of N-nitroso compounds (NOC). Studies with human volunteers showed inhibiting effects of intake of ascorbic acid and green tea consumption on nitrosation using the N-nitrosoproline test. The aim of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service