Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N5154

Sigma-Aldrich

N-Nitrosohexamethyleneimine

Synonym(s):

Hexahydro-1-nitrosoazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O
CAS Number:
Molecular Weight:
128.17
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

liquid

refractive index

n20/D 1.497 (lit.)

bp

233 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=NN1CCCCCC1

InChI

1S/C6H12N2O/c9-7-8-5-3-1-2-4-6-8/h1-6H2

InChI key

UZMVSVHUTOAPTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mutagenic and carcinogenic. A trace-enrichment HPLC method has been developed for quantitating this compound as an impurity (to ppb levels) in tolazamide, and potentially other pharmaceutical preparations.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

210.2 °F - closed cup

flash_point_c

99 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Goodall et al.
Toxicology, 33(3-4), 251-259 (1984-12-01)
The carcinogenic response of NZO/BlGd inbred mice of both sexes to oral N-nitrosohexamethyleneimine (NHEX) was investigated at 2 differing dose rates, but equal cumulative doses. Mice received 52 mg NHEX (2 mg/g body wt) in drinking water over either an
C M Goodall et al.
Journal of the National Cancer Institute, 73(5), 1215-1218 (1984-11-01)
The carcinogenic activity of N-nitrosohexamethyleneimine [(NHEX) CAS: 932-83-2; hexahydro-1-nitroso-1H-azepine] was studied in male and female mice of the four inbred strains NZB/BlGd, NZC/BlGd, NZO/BlGd, and NZY/BlGd. A total of 158 mice received NHEX treatment; 1,338 untreated controls were used, all
L I Hecker et al.
Cancer research, 42(1), 59-64 (1982-01-01)
The in vitro metabolism of N-nitrosohexamethyleneimine by lung and liver microsomes and cytosol from uninduced male Fischer rats is described. Metabolites produced by both organs appeared to be identical. The liver subcellular fractions had a lower Km (0.6 mM) than
A E Ross et al.
Cancer letters, 15(3), 329-334 (1982-03-01)
Liver DNA and RNA were isolated from rats treated with the liver carcinogens N-nitrosopyrrolidine (NPYR) and N-nitrosohexamethyleneimine (NHX). After hydrolysis in 70% perchloric acid (100 degrees C, 1.0 h), 70% of the radioactivity in both the DNA and RNA hydrolysates
G W Harrington et al.
Cancer letters, 32(2), 187-191 (1986-08-01)
Tolazamide, an antidiabetic drug, was found to produce N-nitrosohexamethyleneimine (NHM) upon exposure to an oxidizing agent and in the absence of a nitrosating agent. The oxidizing agents were either hydrogen peroxide or oxygen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service