Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N2251

Sigma-Aldrich

Neotetrazolium chloride form

practical grade, crystalline

Synonym(s):

2,2′,5,5′-Tetraphenyl-3,3′-(p-diphenylene)ditetrazolium chloride, 3,3′-(4,4′-Biphenylene)bis(2,5-diphenyl-2H-tetrazolium chloride), NTC, NeoT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H28Cl2N8
CAS Number:
Molecular Weight:
667.59
Beilstein/REAXYS Number:
3887530
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

practical grade

form

crystalline

color

yellow

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].c1ccc(cc1)-c2nn(-c3ccccc3)[n+](n2)-c4ccc(cc4)-c5ccc(cc5)-[n+]6nc(nn6-c7ccccc7)-c8ccccc8

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xilin Cui et al.
Bioorganic & medicinal chemistry, 16(4), 1927-1947 (2007-12-15)
We have previously reported that sulfated cyclodextrins inhibit the invasion of Plasmodium merozoites by interacting with receptors present on the surface of erythrocytes. The observation that tetrazolium salts formed stable complexes with the inhibitory sulfated cyclodextrins suggested that tetrazolium salts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service