Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N1754

Sigma-Aldrich

1-Nonanol

~98%

Synonym(s):

Alcohol C9, Nonyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8OH
CAS Number:
Molecular Weight:
144.25
Beilstein/REAXYS Number:
969213
EC Number:
MDL number:
UNSPSC Code:
12352200

vapor density

5 (vs air)

vapor pressure

13 mmHg ( 104 °C)

assay

~98%

refractive index

n20/D 1.433 (lit.)

bp

215 °C (lit.)

mp

−8-−6 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCO

InChI

1S/C9H20O/c1-2-3-4-5-6-7-8-9-10/h10H,2-9H2,1H3

InChI key

ZWRUINPWMLAQRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Greene-McDowelle et al.
Toxicon : official journal of the International Society on Toxinology, 37(6), 883-893 (1999-05-26)
The fungi Aspergillus flavus and Aspergillus parasiticus produce the hepatocarcinogenic, secondary metabolites, aflatoxins, in cottonseed, corn, peanuts and treenuts. Results have shown that aflatoxigenic strains of A. flavus and A. parasiticus grown in the presence of specific cotton-leaf volatiles exhibit
M Hori et al.
Journal of pharmaceutical sciences, 81(4), 330-333 (1992-04-01)
The fluxes of representative hydrophilic (propranolol hydrochloride) and lipophilic (diazepam or indomethacin) drugs, administered as ethanolic solutions containing putative penetration enhancers (n-nonane, 1-nonanol, and 1-decanol), were measured across hairless mouse skin in vitro. Propranolol transport was augmented significantly by the
Emmanuel Perisse et al.
BMC biology, 7, 30-30 (2009-06-18)
Synaptic plasticity associated with an important wave of gene transcription and protein synthesis underlies long-term memory processes. Calcium (Ca2+) plays an important role in a variety of neuronal functions and indirect evidence suggests that it may be involved in synaptic
Guanghui Zhao et al.
Journal of hazardous materials, 175(1-3), 715-725 (2009-11-21)
This study investigated the potential of capsules containing 1-nonanol for the adsorption of phenol at high initial concentrations. The polysulfone capsules containing 1-nonanol (PSF@1-nonanol capsules) were successfully prepared with a phase inversion method, and the results showed that 1-nonanol was
Mathias Ditzen et al.
Chemical senses, 28(9), 781-789 (2003-12-05)
The brain's link between perception and action involves several steps, which include stimulus transduction, neuronal coding of the stimulus, comparison to a memory template and choice of an appropriate behavioral response. All of these need time, and many studies report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service