Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N1508

Sigma-Aldrich

N-Nitroso-N-ethylbutylamine

Synonym(s):

Ethylbutylnitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

density

0.91 g/mL (lit.)

SMILES string

CCCCN(CC)N=O

InChI

1S/C6H14N2O/c1-3-5-6-8(4-2)7-9/h3-6H2,1-2H3

InChI key

ZGMCNGHHUQZNIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H U Sons et al.
Cancer, 56(11), 2617-2621 (1985-12-01)
When given N-ethyl-N-butyl-nitrosamine, which was added to the drinking water, a statistically significantly greater number of esophageal papillomas developed in rats with an operatively produced stenosis of the distal esophagus than in control rats without stenosis. The tumor induction occurred
A M Mandard et al.
Bulletin du cancer, 71(5), 419-424 (1984-01-01)
OEsophageal papillomas and carcinomas have been induced in Wistar rats using Ethyl-N-Butyl-Nitrosamine. None of those lesions were observed in a previous experiment on the same rats strains, with ethanol and apple brandy. Experimentally induced tumours in the rat are different
Salah A Sheweita et al.
BioMed research international, 2014, 817019-817019 (2014-09-23)
Millions of people are exposed daily to N-nitrosamines from different environmental sources. The present study aims at investigating the role of N-nitrosamines in the alteration of homocysteine, lipid profile, oxidative stress, paraoxonase activity, antioxidant enzymes, and free radicals which are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service