Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N1258

Sigma-Aldrich

2-Nitroso-1-naphthol

powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ONC10H6OH
CAS Number:
Molecular Weight:
173.17
Colour Index Number:
10010
Beilstein/REAXYS Number:
1867919
EC Number:
MDL number:
UNSPSC Code:
12352103

form

powder

color

yellow

mp

150-155 °C (dec.) (lit.)

SMILES string

Oc1c(ccc2ccccc12)N=O

InChI

1S/C10H7NO2/c12-10-8-4-2-1-3-7(8)5-6-9(10)11-13/h1-6,12H

InChI key

SYUYTOYKQOAVDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bulk package

replaced by

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service