About This Item
refractive index
n20/D 1.495 (lit.)
bp
99-100 °C/6 mmHg (lit.)
density
0.993 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
Cc1cn[nH]c1
InChI
1S/C4H6N2/c1-4-2-5-6-3-4/h2-3H,1H3,(H,5,6)
InChI key
RIKMMFOAQPJVMX-UHFFFAOYSA-N
Gene Information
human ... ADH1A(124) , ADH1B(125) , ADH1C(126)
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
replaced by
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
204.8 °F - closed cup
flash_point_c
96 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service