Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8381

Sigma-Aldrich

N6-Monobutyryl-2′-deoxyadenosine 3′:5′-cyclic monophosphate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H17N5O6PNa
CAS Number:
Molecular Weight:
405.28
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

[Na].CCCC(=O)Nc1ncnc2n(cnc12)C3CC4OP(O)(=O)OCC4O3

InChI

1S/C14H18N5O6P.Na.H/c1-2-3-10(20)18-13-12-14(16-6-15-13)19(7-17-12)11-4-8-9(24-11)5-23-26(21,22)25-8;;/h6-9,11H,2-5H2,1H3,(H,21,22)(H,15,16,18,20);;

InChI key

AXRLNLZXLXWIAP-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service