Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M6656

Sigma-Aldrich

Methyl 15-methylpalmitate

≥98% (capillary GC)

Synonym(s):

15-Methylhexadecanoic acid methyl ester, 15-Methylpalmitic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36O2
CAS Number:
Molecular Weight:
284.48
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

≥98% (capillary GC)

storage temp.

2-8°C

SMILES string

COC(=O)CCCCCCCCCCCCCC(C)C

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service