M6379
2-Methoxyestrone
Synonym(s):
2,3-Dihydroxy-1,3,5(10)-estratrien-17-one 2-methyl ether, 2-Hydroxyestrone 2-methyl ether, 3-Hydroxy-2-methoxy-1,3,5(10)-estratrien-17-one
About This Item
SMILES string
C[C@@]12[C@](CCC2=O)([H])[C@]3([H])CCC4=CC(O)=C(OC)C=C4[C@@]3([H])CC1
Looking for similar products? Visit Product Comparison Guide
related product
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service