Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

M5631

Sigma-Aldrich

MTH-DL-Proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2OS
CAS Number:
Molecular Weight:
170.23
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.26

storage temp.

2-8°C

SMILES string

CN1C(=O)C2CCCN2C1=S

InChI

1S/C7H10N2OS/c1-8-6(10)5-3-2-4-9(5)7(8)11/h5H,2-4H2,1H3

InChI key

NRMNEJAWNNAIBM-UHFFFAOYSA-N

Biochem/physiol Actions

MTH-DL-Proline is a methylthiohydantoin amino acid derivative.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid-Chromatographic separation with Electrochemical Detection of the Phenylthiohydantoin and Methylthiohydantoin Derivatives of Amino-Acids
WELCH LE; JOHNSON DC
Journal of Chromatography A, 180, 111-118 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service