Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4016

Sigma-Aldrich

Methyl methanesulfonate

~99% (GC)

Synonym(s):

Methanesulfonic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3CH3
CAS Number:
Molecular Weight:
110.13
Beilstein/REAXYS Number:
1098586
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

~99% (GC)

refractive index

n20/D 1.414 (lit.)

bp

202-203 °C (lit.)

density

1.3 g/mL at 25 °C (lit.)

SMILES string

COS(C)(=O)=O

InChI

1S/C2H6O3S/c1-5-6(2,3)4/h1-2H3

InChI key

MBABOKRGFJTBAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Principle

DNA adduct; adds methyl groups to DNA at 7-guanine preferentially, but also 3-adenine and 3-guanine.

Other Notes

This product has been replaced by 129925-ALDRICH | Methyl methanesulfonate 99%

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT RE 2 Inhalation

target_organs

Lungs

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

219.2 °F - closed cup

flash_point_c

104 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amaya Azqueta et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(2), 768-773 (2012-12-25)
A serious limitation of the conventional comet assay (single cell gel electrophoresis) is the restriction on the number of samples that can be processed in one experiment, imposed by the size of the electrophoresis platform. One approach to increasing throughput
Isoko Kuriyama et al.
Bioorganic & medicinal chemistry, 21(2), 403-411 (2012-12-26)
The present study was designed to investigate the anticancer activity of novel nine small peptides (compounds 1-9) derived from TT-232, a somatostatin structural analogue, by analyzing the inhibition of mammalian DNA polymerase (pol) and human cancer cell growth. Among the
Cathrine Broberg Vågbø et al.
DNA repair, 12(3), 188-195 (2013-01-02)
Cytotoxic 1-methyladenine (1-meA) and 3-methylcytosine (3-meC) lesions induced in DNA and RNA in vitro and in pre-damaged DNA and RNA bacteriophages in vivo are repaired by the Escherichia coli (E. coli) protein AlkB and a human homolog, ALKBH3. However, it
Stephen Godin et al.
Nucleic acids research, 41(8), 4525-4534 (2013-03-06)
The Saccharomyces cerevisiae Shu complex, consisting of Shu1, Shu2, Csm2 and Psy3, promotes error-free homologous recombination (HR) by an unknown mechanism. Recent structural analysis of two Shu proteins, Csm2 and Psy3, has revealed that these proteins are Rad51 paralogues and
Dongqing Huang et al.
Molecular and cellular biology, 33(8), 1515-1527 (2013-02-06)
Cells employ error-free or error-prone postreplication repair (PRR) processes to tolerate DNA damage. Here, we present a genome-wide screen for sensitivity to 0.001% methyl methanesulfonate (MMS). This relatively low dose is of particular interest because wild-type cells exhibit no discernible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service