Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M3894

Sigma-Aldrich

(R,R)-(+)-N,N′-Bis(α-methylbenzyl)sulfamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H5CH(CH3)NH]2SO2
CAS Number:
Molecular Weight:
304.41
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

mp

98-100 °C (lit.)

SMILES string

C[C@@H](NS(=O)(=O)N[C@H](C)c1ccccc1)c2ccccc2

InChI

1S/C16H20N2O2S/c1-13(15-9-5-3-6-10-15)17-21(19,20)18-14(2)16-11-7-4-8-12-16/h3-14,17-18H,1-2H3/t13-,14-/m1/s1

Inchi Key

ZUMVQOILJDLACR-ZIAGYGMSSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service