Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M2500

Sigma-Aldrich

D-(−)-Mandelic acid

Sigma Grade

Synonym(s):

(R)-α-Hydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O3
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
2691094
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

Sigma Grade

optical activity

[α]20/D ~−155°, c = 2.3 in H2O(lit.)

mp

~133 °C (lit.)

SMILES string

O[C@@H](C(O)=O)c1ccccc1

InChI

1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m1/s1

InChI key

IWYDHOAUDWTVEP-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

The nomenclature of mandelic acid isomers has been confused. This isomer was formerly named L(—)mandelic acid.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

>374.0 °F

flash_point_c

> 190 °C

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer R Bourque et al.
Bioorganic & medicinal chemistry letters, 17(1), 105-108 (2006-10-24)
Mandelate racemase (MR) catalyzes the 1,1-proton transfer that interconverts the enantiomers of mandelate. The transition state/intermediate analogues N-hydroxyformanilide (K(i)=2.79+/-0.19 microM) and cupferron (K(i)=2.67+/-0.09 microM) are identified as potent competitive inhibitors of MR. The pH-pK(i) profile indicates that MR can bind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service