M2022
N,N′-Methylenebisacrylamide
for molecular biology, ≥98%
Synonym(s):
BIS
About This Item
Recommended Products
grade
for molecular biology
assay
≥98%
impurities
<0.1% Acrylic acid content
conductivity
<10 μmho (2% solution)
cation traces
Fe: <40 ppm
Mg: <40 ppm
absorption
≤0.2 at 290 at 1%
foreign activity
DNase, RNase, NICKase and protease, none detected
SMILES string
C=CC(=O)NCNC(=O)C=C
InChI
1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)
InChI key
ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Danger
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral
target_organs
Peripheral nervous system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service