Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M1407

Sigma-Aldrich

Melittin from honey bee venom

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C131H229N39O31
CAS Number:
Molecular Weight:
2846.46
MDL number:
UNSPSC Code:
12352200

assay

≥90% (HPLC)

packaging

vial of 100 μg

storage temp.

2-8°C

SMILES string

CC[C@H](C)[C@H](NC(=O)CN)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc2c[nH]c3ccccc23)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(N)=O)C(N)=O

InChI

1S/C131H229N39O31/c1-23-71(16)102(163-97(176)60-135)122(194)146-62-98(177)148-74(19)109(181)164-100(69(12)13)124(196)160-88(55-65(4)5)116(188)155-84(41-30-33-51-134)115(187)165-101(70(14)15)125(197)161-90(57-67(8)9)118(190)168-106(77(22)173)128(200)169-105(76(21)172)123(195)147-63-99(178)150-92(58-68(10)11)129(201)170-54-36-44-94(170)121(193)149-75(20)108(180)158-89(56-66(6)7)117(189)166-104(73(18)25-3)127(199)162-93(64-171)120(192)159-91(59-78-61-145-80-38-27-26-37-79(78)80)119(191)167-103(72(17)24-2)126(198)157-83(40-29-32-50-133)111(183)154-85(42-34-52-143-130(139)140)112(184)152-82(39-28-31-49-132)110(182)153-86(43-35-53-144-131(141)142)113(185)156-87(46-48-96(137)175)114(186)151-81(107(138)179)45-47-95(136)174/h26-27,37-38,61,65-77,81-94,100-106,145,171-173H,23-25,28-36,39-60,62-64,132-135H2,1-22H3,(H2,136,174)(H2,137,175)(H2,138,179)(H,146,194)(H,147,195)(H,148,177)(H,149,193)(H,150,178)(H,151,186)(H,152,184)(H,153,182)(H,154,183)(H,155,188)(H,156,185)(H,157,198)(H,158,180)(H,159,192)(H,160,196)(H,161,197)(H,162,199)(H,163,176)(H,164,181)(H,165,187)(H,166,189)(H,167,191)(H,168,190)(H,169,200)(H4,139,140,143)(H4,141,142,144)/t71-,72-,73-,74-,75-,76+,77+,81-,82-,83-,84-,85-,86-,87-,88-,89-,90-,91-,92-,93-,94-,100-,101-,102-,103-,104-,105-,106-/m0/s1

InChI key

VDXZNPDIRNWWCW-JFTDCZMZSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-Thr-Thr-Gly-Leu-Pro-Ala-Leu-Ile-Ser-Trp-Ile-Lys-Arg-Lys-Arg-Gln-Gln-NH2

General description

Chemical structure: peptide

Biochem/physiol Actions

Binds calmodulin in a Ca2+-dependent manner; inhibits Na+-K+-ATPase.

Other Notes

The principle hemolytic component of honeybee venom.

Preparation Note

HPLC-purified and use tested as a protease substrate.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua L Hood et al.
Antiviral therapy, 18(1), 95-103 (2012-09-08)
We investigated whether cytolytic melittin peptides could inhibit HIV-1 infectivity when carried in a nanoparticle construct that might be used as a topical vaginal virucide. Free melittin and melittin-loaded nanoparticles were prepared and compared for cytotoxicity and their ability to
Kirk K Hou et al.
Biomaterials, 34(12), 3110-3119 (2013-02-06)
Traditional transfection agents including cationic lipids and polymers have high efficiency but cause cytotoxicity. While cell penetrating peptide based transfection agents exhibit improved cytotoxicity profiles, they do not have the efficiency of existing lipidic agents due to endosomal trapping. As
C E Dempsey
Biochimica et biophysica acta, 1031(2), 143-161 (1990-05-07)
The molecular mechanisms underlying the various effects of melittin on membranes have not been completely defined and much of the evidence described indicates that different molecular mechanisms may underlie different actions of the peptide. Ideas about the formation of transbilayer
Babak Y Moghadam et al.
Langmuir : the ACS journal of surfaces and colloids, 28(47), 16318-16326 (2012-08-28)
Lipid bilayers are biomembranes common to cellular life and constitute a continuous barrier between cells and their environment. Understanding the interaction of engineered nanomaterials (ENMs) with lipid bilayers is an important step toward predicting subsequent biological effects. In this study
Alberto Falco et al.
Antiviral research, 97(2), 218-221 (2012-12-25)
In this study, melittin, a well-characterized pore-forming lytic amphiphilic peptide susceptible to be vehiculized in lipid membranes, has been utilized to study their antiviral properties. For this purpose, an assay based on melittin loaded-immunoliposomes previously described by our group was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service