Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M121

Sigma-Aldrich

(−)-MDO-NPA hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO2 · xHCl
CAS Number:
Molecular Weight:
307.39 (free base basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

mp

250-252 °C

SMILES string

Cl.CCCN1CCc2cccc-3c2[C@H]1Cc4ccc5OCOc5c-34

InChI

1S/C20H21NO2.ClH/c1-2-9-21-10-8-13-4-3-5-15-18(13)16(21)11-14-6-7-17-20(19(14)15)23-12-22-17;/h3-7,16H,2,8-12H2,1H3;1H/t16-;/m1./s1

InChI key

QRTFNNCNSYALRJ-PKLMIRHRSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An orally effective, long-acting dopaminergic prodrug: (-)-10,11-methylenedioxy-N-propylnoraporphine.
R J Baldessarini et al.
European journal of pharmacology, 77(1), 87-88 (1982-01-08)
P Lampen et al.
Journal of chromatography, 426(2), 283-294 (1988-04-29)
The dopamine receptor agonist R(-)N-n-propylnorapomorphine (NPA) and its proposed pro-drug R(-)10,11-methylenedioxy-N-n-propylnoraporphine (MDO-NPA) were isolated simultaneously from monkey plasma using a solid-phase extraction procedure. R(-)Apomorphine (APO) and R(-)10,11-methylenedioxyaporphine (MDO-APO) were added as internal standards, and separation and quantification were by high-performance
A Campbell et al.
Brain research, 403(2), 393-397 (1987-02-17)
The 10-11-methylenedioxy (MDO) derivative of S(+)N-n-propylnorapomorphine (NPA) was prepared and tested as a possible active prodrug to S(+)NPA, which we have recently found to exert in vivo activity suggestive of selective antagonism of dopamine receptors in the limbic forebrain but
A Campbell et al.
Neuropharmacology, 21(10), 953-961 (1982-10-01)
Substituted and unsubstituted 10,11-methylenedioxy derivatives of apomorphine (APO) or its N-propyl congener (NPA) were synthesized and evaluated for their ability to alter motor activity or to induce stereotyped behavior in the rat. Of these, (-)10,11-methylenedioxy-N-n-propylnoraporphine hydrochloride (MDO-NPA) was the most
(-)-10,11-Methylenedioxy-N-propylnoraporphine, an orally effective dopamine agonist and duodenal antiulcerogen in the rat.
J L Neumeyer et al.
European journal of pharmacology, 88(2-3), 273-274 (1983-03-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service