About This Item
form
aqueous ethanol solution
extent of labeling
15-30 Ci/mmol
mass shift
M+4
storage temp.
2-8°C
SMILES string
[3H]C([3H])(O)[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
Packaging
Physical form
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
68.0 °F - closed cup
flash_point_c
20 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service