Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L9024

Sigma-Aldrich

Leukotriene B5

~100 μg/mL in ethanol, ≥97% (HPLC)

Synonym(s):

([5S,12R]-Dihydroxy-[6Z,8E,10E,14Z,17Z]-eicosapentaenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O4
CAS Number:
Molecular Weight:
334.45
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

≥97% (HPLC)

concentration

~100 μg/mL in ethanol

shipped in

dry ice

storage temp.

−70°C

SMILES string

CC\C=C\C\C=C\CC(O)\C=C\C=C\C=C\C(O)CCCC(O)=O

Packaging

Packaged under argon.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akifumi Hagi et al.
JPEN. Journal of parenteral and enteral nutrition, 34(3), 263-270 (2010-05-15)
This study investigated the effects of parenterally administered fish oil (FO) on the fatty acid composition in rats to determine the optimal omega-6:omega-3 polyunsaturated fatty acid (PUFA) ratio of fat emulsions to achieve an anti-inflammatory effect. Male Sprague-Dawley rats were
M K Boudreaux et al.
Journal of the American Animal Hospital Association, 33(3), 235-243 (1997-05-01)
Thirty beagles were placed on diets containing ratios of n-6 to n-3 fatty acids ranging from 5:1 to 100:1 for 12 weeks to determine the effects of these diets on platelet reactivity, coagulation screening assays, and antithrombin III activity. Although
N Nakamura et al.
The Journal of clinical investigation, 92(3), 1253-1261 (1993-09-01)
Leukotriene (LT) B4 is a major chemical activator of PMN. Inhibitory effects of oral administration of docosahexaenoic acid (DHA) on LTB4 synthesis by PMN are known. We intravenously infused tridocosahexaenoyl-glycerol (DHA-TG) emulsion into rabbits in three different doses, namely 0.8
N Matsuo et al.
Bioscience, biotechnology, and biochemistry, 64(7), 1437-1443 (2000-08-17)
Male Wistar rats were given purified diets containing safflower (SAF), perilla (PER), or palm (PAL) oils with or without 1% tea polyphenols (TP) for 3 weeks, and chemical mediator releasing activity from rat peritoneal exudate cells (PEC) was measured. Histamine
The effect of n-3 fatty acids on leukotriene formation from neutrophils in patients on hemodialysis.
K Løssl et al.
Lipids, 34 Suppl, S185-S185 (1999-07-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service