Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K2139

Sigma-Aldrich

K-252b solution

≥98% (HPLC), from Nonomuraea longicatena, ready-to-use solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H19N3O5
CAS Number:
Molecular Weight:
453.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

Nonomuraea longicatena

assay

≥98% (HPLC)

form

ready-to-use solution

concentration

1 mM in DMSO

shipped in

wet ice

storage temp.

−20°C

SMILES string

[H][C@]1(C[C@]2(O)C(O)=O)O[C@]2(C)N3C4=C(C=CC=C4)C5=C3C(N1C6=C7C=CC=C6)=C7C8=C5CNC8=O

InChI

1S/C26H19N3O5/c1-25-26(33,24(31)32)10-17(34-25)28-15-8-4-2-6-12(15)19-20-14(11-27-23(20)30)18-13-7-3-5-9-16(13)29(25)22(18)21(19)28/h2-9,17,33H,10-11H2,1H3,(H,27,30)(H,31,32)/t17-,25+,26+/m1/s1

InChI key

AMSOPBXQXSAAAC-PLZPTFKGSA-N

Biochem/physiol Actions

Non-selective protein kinase inhibitor; ecto-protein kinase inhibitor; PKC inhibitor

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

188.6 °F

flash_point_c

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fujii et al.
Neuroscience, 113(3), 617-628 (2002-08-02)
The mechanism of ATP-induced long-term potentiation (LTP) was studied pharmacologically using guinea-pig hippocampal slices. LTP, induced in CA1 neurons by 10 min application of 10 microM ATP, was blocked by co-application of the N-methyl-D-aspartate (NMDA) receptor antagonist, D,L-2-amino-5-phosphonovalerate (5 or
Luis Cartuche et al.
Biomolecules, 10(4) (2020-04-30)
Chagas disease and leishmaniasis are neglected tropical diseases caused by kinetoplastid parasites of Trypanosoma and Leishmania genera that affect poor and remote populations in developing countries. These parasites share similar complex life cycles and modes of infection. It has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service