Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K1382

Sigma-Aldrich

Kojibiose

≥95% (HPAE/PAD)

Synonym(s):

α-D-Glc-(1→2)-D-Glc, 2-O-α-D-Glucopyranosyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

assay

≥95% (HPAE/PAD)

storage temp.

−20°C

SMILES string

OCC(O)C(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C=O

InChI

1S/C12H22O11/c13-1-4(16)7(17)8(18)5(2-14)22-12-11(21)10(20)9(19)6(3-15)23-12/h2,4-13,15-21H,1,3H2

InChI key

PZDOWFGHCNHPQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Kojibiose is an inhibitor of plant glucosidase I. It inhibits the removal of terminal glucose from (Glc)3(Man)9(GlcNAc)2.
Kojibiose is an inhibitor of plant glucosidase I. It inhibits the removal of terminal glucose from the high-mannose oligosaccharide (Glc)3(Man)9(GlcNAc)2, either from the free oligosaccharide or from the oligosaccharide attached to a protein via N-linkage.

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min-Sun Kang et al.
Biochimie, 91(11-12), 1434-1442 (2009-08-18)
Genome analysis of Lactobacillus johnsonii NCC533 has been recently completed. One of its annotated genes, lj0569, encodes the protein having the conserved domain of glycoside hydrolase family 31. Its homolog gene (ljag31) in L. johnsonii NBRC13952 was cloned and expressed
Wouter F J Hogendorf et al.
Bioorganic & medicinal chemistry, 18(11), 3668-3678 (2010-04-23)
In this paper the synthesis of an Enterococcus Faecalis teichoic acid (TA) hexamer is presented. The key kojibiosyl-glycerol phosphoramidite building block was obtained by condensation of thioglucose donors, provided with various protecting groups at the C2 hydroxyl function with an
Takuo Yamamoto et al.
Bioscience, biotechnology, and biochemistry, 75(6), 1208-1210 (2011-06-15)
One kojibiose phoshorylase (KP) homolog gene was cloned from Caldicellulosiruptor saccharolyticus ATCC43494. Recombinant KP from C. saccharolyticus (Cs-KP) expressed in Escherichia coli showed highest activity at pH 6.0 at 85 °C, and was stable from pH 3.5 to 10.0 and
Takuo Yamamoto et al.
Journal of bioscience and bioengineering, 100(3), 343-346 (2005-10-26)
The kojibiose phosphorylase (KP) gene and trehalose phosphorylase (TP) gene from Thermoanaerobacter brockii ATCC35047 were intracellularly hyper-expressed under the control of the Bacillus amyloliquefaciens alpha-amylase promoter in Bacillus subtilis. The production yields were estimated to be 2.1 g of KP
A R Santa Cruz et al.
Experientia, 41(7), 928-929 (1985-07-15)
We have prepared dolichylpyrophosphoryl-[14C]-oligosaccharide (Dol-PP-oligosaccharide) from calf thyroid. Microsomal fractions from human breast tissues catalyzed the transfer of labeled oligosaccharide to endogenous acceptor proteins. Malignant tumors showed higher activity of the oligosaccharide transferring enzyme than normal tissue. With kojibiose (Kj)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service