Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I7772

SAFC

Sodium ionophore VI

Synonym(s):

Bis[(12-crown-4)methyl] dodecylmethylmalonate, Dodecylmethylmalonic acid bis[(12-crown-4)methyl ester]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H62O12
CAS Number:
Molecular Weight:
662.85
Beilstein/REAXYS Number:
4221107
MDL number:
UNSPSC Code:
12352200

storage temp.

2-8°C
2-8°C

SMILES string

CCCCCCCCCCCCC(C)(C(=O)OCC1COCCOCCOCCO1)C(=O)OCC2COCCOCCOCCO2

InChI

1S/C34H62O12/c1-3-4-5-6-7-8-9-10-11-12-13-34(2,32(35)45-28-30-26-41-20-18-37-14-16-39-22-24-43-30)33(36)46-29-31-27-42-21-19-38-15-17-40-23-25-44-31/h30-31H,3-29H2,1-2H3

InChI key

FJSZDECKJYYBGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Macrocyclic polyethers with repeating (-CH2CH2O) units. The compounds are ionophoric (form stable complexes with cations).

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Schueler et al.
PloS one, 6(11), e27167-e27167 (2011-11-15)
Transport metabolons have been discussed between carbonic anhydrase II (CAII) and several membrane transporters. We have now studied different CA isoforms, expressed in Xenopus oocytes alone and together with the electrogenic sodium bicarbonate cotransporter 1 (NBCe1), to determine their catalytic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service