Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I5377

SAFC

Propham

~99%, crystalline

Synonym(s):

IPC, Isopropyl phenylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
Beilstein/REAXYS Number:
2209666
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

~99%

form

crystalline

color

tan

SMILES string

CC(C)OC(=O)Nc1ccccc1

InChI

1S/C10H13NO2/c1-8(2)13-10(12)11-9-6-4-3-5-7-9/h3-8H,1-2H3,(H,11,12)

InChI key

VXPLXMJHHKHSOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

One gram gratis to non-profit institutions. Repeat requests are welcomed.

comparable product

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Surcel et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(5), 1428-1433 (2015-01-22)
Current approaches to cancer treatment focus on targeting signal transduction pathways. Here, we develop an alternative system for targeting cell mechanics for the discovery of novel therapeutics. We designed a live-cell, high-throughput chemical screen to identify mechanical modulators. We characterized
Salvatore Piscuoglio et al.
Molecular oncology, 8(8), 1588-1602 (2014-07-22)
Papillary carcinoma (PC) is a rare type of breast cancer, which comprises three histologic subtypes: encapsulated PC (EPC), solid PC (SPC) and invasive PC (IPC). Microarray-based gene expression and Affymetrix SNP 6.0 gene copy number profiling, and RNA-sequencing revealed that
Yael Weiss et al.
Nature protocols, 1(5), 2282-2287 (2007-04-05)
Herbicide-resistant plants can be generated by either traditional breeding procedures or genetic engineering. Analyses of plant responses to a newly developed herbicide or the tolerance level of a newly developed plant line to a given herbicide are based on various
B Caporiccio et al.
Comptes rendus des seances de la Societe de biologie et de ses filiales, 175(4), 496-500 (1981-01-01)
The bibliographic data dealing with the acute toxicity of two phenylcarbamates, propham and chloropropham, are not very precise an often contradictory. This work is a restatement and also a demonstration of the high sensitivity of the aquatic animals that we
D G Hoover et al.
Applied and environmental microbiology, 51(2), 226-232 (1986-02-01)
The rates of mineralization of nitrilotriacetic acid (NTA), 2,4-dichlorophenoxyacetic acid (2,4-D), p-nitrophenol, aniline, and isopropyl N-phenylcarbamate (IPC) at one or more concentrations ranging from 100 pg/ml to 1.0 microgram/ml were proportional to chemical concentrations in samples of three lakes. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service