Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I3011

Sigma-Aldrich

myo-Inositol

suitable for plant cell culture

Synonym(s):

1,2,3,4,5,6-Hexahydroxycyclohexane, i-Inositol, meso-Inositol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
1907329
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

vapor density

6.2 (vs air)

technique(s)

cell culture | plant: suitable

mp

222-227 °C (lit.)

SMILES string

O[C@H]1[C@H](O)[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4+,5-,6-

InChI key

CDAISMWEOUEBRE-GPIVLXJGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Utriainen et al.
Journal of neuro-oncology, 62(3), 329-338 (2003-06-05)
The signal of choline containing compounds (Cho) in proton magnetic resonance spectroscopy (1H-MRS) is elevated in brain tumors. [11C]choline uptake as assessed using positron emission tomography (PET) has also been suggested to be higher in brain tumors than in the
Pascal M Groenen et al.
American journal of obstetrics and gynecology, 189(6), 1713-1719 (2004-01-08)
The purpose of this study was to investigate the maternal and children's myo-inositol, glucose, and zinc status in association with spina bifida risk. Sixty-three mothers and 70 children with spina bifida and 102 control mothers and 85 control children were
C Servo et al.
Diabetologia, 11(6), 575-580 (1975-12-01)
Cerebrospinal fluid (CSF) or CSF and plasma levels of sorbitol, 1,5-anhydroglucitol and myoinositol of diabetic and non-diabetic patients with normal kidney function and of diabetic and non-diabetic patients with impaired kidney function were measured by gas-liquid chromatography. The CSF sorbitol
Nathan Czyzewicz et al.
Journal of experimental botany, 66(17), 5229-5243 (2015-07-19)
Plant roots are important for a wide range of processes, including nutrient and water uptake, anchoring and mechanical support, storage functions, and as the major interface with the soil environment. Several small signalling peptides and receptor kinases have been shown
Nathan Czyzewicz et al.
Journal of experimental botany, 66(17), 5367-5374 (2015-07-03)
In the Arabidopsis thaliana genome, over 1000 putative genes encoding small, presumably secreted, signalling peptides can be recognized. However, a major obstacle in identifying the function of genes encoding small signalling peptides is the limited number of available loss-of-function mutants.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service