Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I1631

Sigma-Aldrich

4-Imino-1,3-diazabicyclo[3.1.0]hexan-2-one

~99% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3O
CAS Number:
Molecular Weight:
111.10
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

~99% (TLC)

storage temp.

2-8°C

SMILES string

N=C1NC(=O)N2CC12

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service