Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H9754

Sigma-Aldrich

Heneicosane

~99%, crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)19CH3
CAS Number:
Molecular Weight:
296.57
Beilstein/REAXYS Number:
1748500
MDL number:
UNSPSC Code:
12352100

vapor density

10.3 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

assay

~99%

form

crystalline

bp

100 °C/2 mmHg (lit.)
356.1 °C/760 mmHg

mp

39-41 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCC

InChI

1S/C21H44/c1-3-5-7-9-11-13-15-17-19-21-20-18-16-14-12-10-8-6-4-2/h3-21H2,1-2H3

InChI key

FNAZRRHPUDJQCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service