Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H9654

Sigma-Aldrich

1′-Hydroxy-2′-acetonaphthone

Synonym(s):

2-Acetyl-1-naphthol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H6COCH3
CAS Number:
Molecular Weight:
186.21
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

98-100 °C (lit.)

SMILES string

CC(=O)c1ccc2ccccc2c1O

InChI

1S/C12H10O2/c1-8(13)10-7-6-9-4-2-3-5-11(9)12(10)14/h2-7,14H,1H3

Inchi Key

JBGJVMVWYWUVOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gamze Bora-Tatar et al.
Bioorganic & medicinal chemistry, 17(14), 5219-5228 (2009-06-13)
In the light of known HDAC inhibitors, 33 carboxylic acid derivatives were tested to understand the structural requirements for HDAC inhibition activity. Several modifications were applied to develop the structure-activity relationships of carboxylic acid HDAC inhibitors. HDAC inhibition activities were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service