Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H9646

Sigma-Aldrich

1-O-Palmityl-2-acetyl-rac-glycerol

~95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H42O4
CAS Number:
Molecular Weight:
358.56
MDL number:
UNSPSC Code:
12352211

assay

~95%

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCOCC(CO)OC(C)=O

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service