Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H6153

Sigma-Aldrich

gp120 Fragment 421-438

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C99H148N24O25S2
CAS Number:
Molecular Weight:
2138.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥90% (HPLC)

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](N)CCCCN)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](Cc2c[nH]c3ccccc23)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](Cc4ccc(O)cc4)C(=O)N[C@@H](C)C(=O)N5CCC[C@H]5C(=O)N6CCC[C@H]6C(O)=O

InChI

1S/C99H148N24O25S2/c1-9-54(4)82(121-94(142)71(47-57-21-11-10-12-22-57)117-87(135)65(33-36-76(103)125)112-84(132)62(102)24-15-17-41-100)96(144)119-73(50-78(105)127)93(141)115-69(40-46-150-8)89(137)118-72(49-59-51-106-63-25-14-13-23-61(59)63)92(140)114-66(34-37-77(104)126)86(134)113-67(35-38-80(129)130)90(138)120-81(53(2)3)95(143)107-52-79(128)110-64(26-16-18-42-101)85(133)108-55(5)83(131)111-68(39-45-149-7)88(136)116-70(48-58-29-31-60(124)32-30-58)91(139)109-56(6)97(145)122-43-19-27-74(122)98(146)123-44-20-28-75(123)99(147)148/h10-14,21-23,25,29-32,51,53-56,62,64-75,81-82,106,124H,9,15-20,24,26-28,33-50,52,100-102H2,1-8H3,(H2,103,125)(H2,104,126)(H2,105,127)(H,107,143)(H,108,133)(H,109,139)(H,110,128)(H,111,131)(H,112,132)(H,113,134)(H,114,140)(H,115,141)(H,116,136)(H,117,135)(H,118,137)(H,119,144)(H,120,138)(H,121,142)(H,129,130)(H,147,148)/t54-,55-,56-,62-,64-,65-,66-,67-,68-,69-,70-,71-,72-,73-,74-,75-,81-,82-/m0/s1

InChI key

FWLUYNSJWIJPKZ-WEARTWMDSA-N

Amino Acid Sequence

Lys-Gln-Phe-Ile-Asn-Met-Trp-Gln-Glu-Val-Gly-Lys-Ala-Met-Tyr-Ala-Pro-Pro

General description

The gp120 (glycoprotein-120) is a HIV (human immunodeficiency virus)-1 envelope glycoprotein. The entry of the virus in cells requires association between gp120 and cell surface receptors, CD4 (cluster of differentiation 4) and chemokine receptor.[1][2]

Other Notes

Peptide derived from the CD4 attachment region of HIV gp120.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W J Morrow et al.
Immunology, 75(4), 557-564 (1992-04-01)
In our efforts to identify products that might be used for active immunotherapy in human immunodeficiency virus (HIV) infection, we have studied synthetic peptides derived from the CD4 attachment site of gp120. Two peptides have emerged with particularly interesting properties.
R Wyatt et al.
Nature, 393(6686), 705-711 (1998-06-26)
The human immunodeficiency virus HIV-1 establishes persistent infections in humans which lead to acquired immunodeficiency syndrome (AIDS). The HIV-1 envelope glycoproteins, gp120 and gp41, are assembled into a trimeric complex that mediates virus entry into target cells. HIV-1 entry depends
P D Kwong et al.
Nature, 393(6686), 648-659 (1998-06-26)
The entry of human immunodeficiency virus (HIV) into cells requires the sequential interaction of the viral exterior envelope glycoprotein, gp120, with the CD4 glycoprotein and a chemokine receptor on the cell surface. These interactions initiate a fusion of the viral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service