Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H5885

Supelco

Hydrocortisone

Sigma Reference Standard

Synonym(s):

11β,17α,21-Trihydroxypregn-4-ene-3,20-dione, 17-Hydroxycorticosterone, 4-Pregnene-11β,17α,21-triol-3,20-dione, Cortisol, Kendall’s compound F, Reichstein’s substance M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O5
CAS Number:
Molecular Weight:
362.46
Beilstein/REAXYS Number:
1354819
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

Sigma Reference Standard

packaging

vial of 250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

211-214 °C (lit.)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

JYGXADMDTFJGBT-VWUMJDOOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Primary glucocorticoid secreted by the adrenal cortex.
Primary glucocorticoid secreted by the adrenal cortex. It has three times the anti-inflammatory potency of corticosterone but much lower Na2+ retention potency.

Packaging

Supplied in amber screw-cap vials

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Repr. 1A - STOT RE 2

target_organs

Adrenal gland,Bone marrow

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liselotte van Bloemendaal et al.
Diabetes, 63(12), 4186-4196 (2014-07-30)
Gut-derived hormones, such as GLP-1, have been proposed to relay information to the brain to regulate appetite. GLP-1 receptor agonists, currently used for the treatment of type 2 diabetes (T2DM), improve glycemic control and stimulate satiety, leading to decreases in
K J Rademaker et al.
Archives of disease in childhood. Fetal and neonatal edition, 93(1), F58-F63 (2007-09-13)
The benefits versus the risks of postnatal administration of steroids in preterm-born infants are still debatable. This review examines the literature on postnatal hydrocortisone treatment for chronic lung disease (CLD) in preterm-born infants with a particular focus on the effects
Florian Hardeveld et al.
Psychoneuroendocrinology, 50, 62-71 (2014-09-03)
Although HPA-axis activity has been studied extensively in relation to depression, there is no consensus whether HPA-axis parameters predicts major depressive disorder (MDD) recurrence. We investigated whether HPA-axis parameters (cortisol awakening response (CAR), the dexamethasone suppression test (DST) and evening
Lorenz Goette et al.
Psychoneuroendocrinology, 54, 115-123 (2015-02-24)
Social competition is a fundamental mechanism of evolution and plays a central role in structuring individual interactions and communities. Little is known about the factors that affect individuals' competitive success, particularly in humans. Key factors might include stress, a major
Rachel Yehuda et al.
The American journal of psychiatry, 171(8), 872-880 (2014-05-17)
Differential effects of maternal and paternal posttraumatic stress disorder (PTSD) have been observed in adult offspring of Holocaust survivors in both glucocorticoid receptor sensitivity and vulnerability to psychiatric disorder. The authors examined the relative influences of maternal and paternal PTSD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service