H3764
meso-Hydrobenzoin
Synonym(s):
meso-1,2-Diphenyl-1,2-ethanediol
About This Item
Recommended Products
mp
137-139 °C (lit.)
storage temp.
2-8°C
SMILES string
O[C@H]([C@H](O)c1ccccc1)c2ccccc2
InChI
1S/C14H14O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-16H/t13-,14+
InChI key
IHPDTPWNFBQHEB-OKILXGFUSA-N
Looking for similar products? Visit Product Comparison Guide
Application
replaced by
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service