H3635
γ-Hydroxybutyric acid sodium salt
Synonym(s):
4-Hydroxybutyric acid sodium salt, γ-Hydroxybutyric acid sodium salt, GHB, Sodium 4-hydroxybutyrate
About This Item
Recommended Products
drug control
USDEA Schedule I; Home Office Schedule 4.1; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
SMILES string
[Na+].OCCCC([O-])=O
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service